missing translation for 'onlineSavingsMsg'
Learn More

Tiotropium Bromide 98.0+%, TCI America™

Catalog No. p-7137538
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T303250MG 50 mg
T3032200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T303250MG Supplier TCI America Supplier No. T303250MG

Chemical Identifiers

CAS 136310-93-5
Molecular Formula C19H22BrNO4S2
Molecular Weight (g/mol) 472.412
MDL Number MFCD00867027
InChI Key DQHNAVOVODVIMG-RGECMCKFSA-M
PubChem CID 5487426
SMILES C[N+]1(C2CC(CC1C3C2O3)OC(=O)C(C4=CC=CS4)(C5=CC=CS5)O)C.[Br-]

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 50 mg
Formula Weight 472.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tiotropium Bromide