missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tiopronin | |
| 98°C | |
| C5H9NO3S | |
| 5 g | |
| YTGJWQPHMWSCST-UHFFFAOYNA-N | |
| 2-(2-sulfanylpropanoylamino)acetic acid | |
| 5483 | |
| 163.19 | |
| Crystalline Powder |
| 2-2-1953 | |
| White-Yellow | |
| MFCD00004861 | |
| N-(2-Mercaptopropionyl)glycine | |
| CC(C(=O)NCC(=O)O)S | |
| 163.19 | |
| CHEBI:32229 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 2-2-1953 | |
| 163.19 | |
| YTGJWQPHMWSCST-UHFFFAOYNA-N | |
| 5483 | |
| 2-(2-sulfanylpropanoylamino)acetic acid |
| C5H9NO3S | |
| MFCD00004861 | |
| N-(2-Mercaptopropionyl)glycine | |
| CHEBI:32229 | |
| CC(C(=O)NCC(=O)O)S |
Safety and Handling
EINECSNumber : (2)-1207&(9)-1423
RTECSNumber : MC0596500
TSCA : No