missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiopronin 97.0+%, TCI America™
$207.92 - $539.05
Chemical Identifiers
| CAS | 2-2-1953 |
|---|---|
| Molecular Formula | C5H9NO3S |
| Molecular Weight (g/mol) | 163.19 |
| MDL Number | MFCD00004861 |
| InChI Key | YTGJWQPHMWSCST-UHFFFAOYNA-N |
| Synonym | N-(2-Mercaptopropionyl)glycine |
| PubChem CID | 5483 |
| ChEBI | CHEBI:32229 |
| IUPAC Name | 2-(2-sulfanylpropanoylamino)acetic acid |
| SMILES | CC(C(=O)NCC(=O)O)S |
Chemical Identifiers
| 2-2-1953 | |
| 163.19 | |
| YTGJWQPHMWSCST-UHFFFAOYNA-N | |
| 5483 | |
| 2-(2-sulfanylpropanoylamino)acetic acid |
| C5H9NO3S | |
| MFCD00004861 | |
| N-(2-Mercaptopropionyl)glycine | |
| CHEBI:32229 | |
| CC(C(=O)NCC(=O)O)S |
Specifications
| 2-2-1953 | |
| White-Yellow | |
| MFCD00004861 | |
| N-(2-Mercaptopropionyl)glycine | |
| CC(C(=O)NCC(=O)O)S | |
| 163.19 | |
| CHEBI:32229 | |
| ≥97.0% (GC,T) | |
| Tiopronin |
| 98°C | |
| C5H9NO3S | |
| 5 g | |
| YTGJWQPHMWSCST-UHFFFAOYNA-N | |
| 2-(2-sulfanylpropanoylamino)acetic acid | |
| 5483 | |
| 163.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1207&(9)-1423
RTECSNumber : MC0596500
TSCA : No