missing translation for 'onlineSavingsMsg'
Learn More

Tioconazole Impurity Standard, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022918
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022918 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022918 Supplier Merck Emd Millipore Supplier No. BP687

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 65899-73-2
Molecular Formula C16H13Cl3N2OS
Linear Formula C16H13Cl3N2OS
MDL Number MFCD00057276
Synonym Tioconazole; 1-[2-[(2-Chloro-3-thienyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1 H-imidazole; Thioconazole
Type Pharmacopeia and Metrological Institutes Standards
Quantity 100 mg
Formula Weight 387.71
Grade British Pharmacopoeia (BP) Reference Standard