missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin Standard For ICP, 1000μg/mL in 20% HCl, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$385.35 - $634.88
Chemical Identifiers
| CAS | 7440-31-5 |
|---|---|
| Molecular Formula | Sn |
| Molecular Weight (g/mol) | 118.71 |
| MDL Number | MFCD00133862 |
| InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
| PubChem CID | 5352426 |
| ChEBI | CHEBI:32990 |
| IUPAC Name | tin |
| SMILES | [Sn] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Sn | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-31-5 | |
| 118.71 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| CHEBI:32990 | |
| [Sn] |
| Sn | |
| MFCD00133862 | |
| 5352426 | |
| tin |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 1.0,20.0,79.0 | |
| Sn | |
| AA, ICP | |
| Certified Reference Material | |
| [Sn] | |
| tin | |
| 125 mL | |
| CHEBI:32990 | |
| Matrix Concentration Cat. No. 1% HNO3/1% HF 1000μg/mL PLSN2-2X 2% HNO3/2% HF 10,000 PLSN2-3X 20% HCl 1000μg/mL PLSN5-2Y, -2T, -2X 10,000 PLSN5-3Y, -3X |
| 7440-31-5,7647-01-0,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 1.0,20.0,79.0 | |
| MFCD00133862 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| 20% HCl | |
| 118.71 | |
| 5352426 | |
| Assurance | |
| Tin Standard |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only