missing translation for 'onlineSavingsMsg'
Learn More

Tin selenide, 99.999% (metals basis)

CAS: 1315-06-6 | H2SeSn | 199.70 g/mol

$236.55 - $824.40

Chemical Identifiers

CAS 1315-06-6
Molecular Formula SnSe
Molecular Weight (g/mol) 197.65
MDL Number MFCD00054049
InChI Key MFIWAIVSOUGHLI-UHFFFAOYSA-N
IUPAC Name selenoxo-lambda~2~-stannane
SMILES [Se]=[SnH2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1878106
View Documents
Thermo Scientific Chemicals
01878106
5 g
Each for $236.55
Only null left
 
AA1878114
View Documents
Thermo Scientific Chemicals
01878114
25 g
Each for $824.40
Only null left
 
Description

Description

Tin selenide finds application in low- cost photovoltaics devices and memory-switching devices. It acts as a solid-state lubricant. It is also used for solar cells and anodes for lithium-ion batteries. Further, it is a narrow band-gap semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1315-06-6
197.65
MFIWAIVSOUGHLI-UHFFFAOYSA-N
[Se]=[SnH2]
SnSe
MFCD00054049
selenoxo-lambda~2~-stannane

Specifications

861°C
Pieces
99.999% (metals basis)
MFCD00054049
MFIWAIVSOUGHLI-UHFFFAOYSA-N
selenoxo-lambda~2~-stannane
197.65
1315-06-6
5 g
SnSe
Insoluble in water. Soluble in aqua regia,alkali sulfides,and alkali selenides.
[Se]=[SnH2]
197.65
Tin selenide
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 215-257-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only