accessibility menu, dialog, popup

UserName

H2SnSe

CAS RN 1315-06-6

IUPAC Name: selenoxo-lambda~2~-stannane
Synonyms: tin selenide
Molecular Weight (g/mol): 197.65
Molecular Formula: SnSe
InChi Key: MFIWAIVSOUGHLI-UHFFFAOYSA-N
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results