missing translation for 'onlineSavingsMsg'
Learn More

Tin Metal, Spectrum™ Chemical

$395.18 - $3823.68

Chemical Identifiers

CAS 7440-31-5
Molecular Weight (g/mol) 118.71
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614445
View Documents
Spectrum Chemical Mfg Cor
T1061100GM
100 g Poly Bottle
Each for $395.18
Only null left
 
18614451
View Documents
Spectrum Chemical Mfg Cor
T1061500GM
500 g Poly Bottle
Each for $1,024.21
Only null left
 
18614446
View Documents
Spectrum Chemical Mfg Cor
T106125KG
2.5 kg Poly Bottle
Each for $3,823.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7440-31-5
118.71

Specifications

7440-31-5
100 g
Powder
Ungraded
1
Poly Bottle
118.71
Tin Metal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.