missing translation for 'onlineSavingsMsg'
Learn More

Tin Metal, Spectrum™ Chemical

Catalog No. 18614446
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
2.5 kg
500 g
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
18614446 2.5 kg Poly Bottle
18614445 100 g Poly Bottle
18614451 500 g Poly Bottle
3 options

Catalog No. 18614446

Supplier: Spectrum Chemical Mfg Cor T10612.5KG

Only null left

Chemical Identifiers

CAS 7440-31-5
Molecular Weight (g/mol) 118.71

Specifications

CAS Max % 1
Quantity 2.5 kg
Packaging Poly Bottle
Form Powder
Grade Ungraded
Chemical Name or Material Tin Metal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.