missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) Chloride Pentahydrate, 98%, Honeywell Fluka™
>98%
$1105.73 - $1105.73
Chemical Identifiers
| CAS | 10026-06-9 |
|---|---|
| Molecular Formula | Cl4H10O5Sn |
| Molecular Weight (g/mol) | 350.59 |
| MDL Number | MFCD00149864 |
| InChI Key | KHMOASUYFVRATF-UHFFFAOYSA-J |
| Synonym | tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o |
| PubChem CID | 61442 |
| IUPAC Name | tin(4+) pentahydrate tetrachloride |
| SMILES | O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 10026-06-9 | |
| 350.59 | |
| KHMOASUYFVRATF-UHFFFAOYSA-J | |
| 61442 | |
| O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4] |
| Cl4H10O5Sn | |
| MFCD00149864 | |
| tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o | |
| tin(4+) pentahydrate tetrachloride |
Specifications
| White to Yellow | |
| 1 kg | |
| SnCl4 · 5H2O | |
| UN2440 | |
| KHMOASUYFVRATF-UHFFFAOYSA-J | |
| tin(4+) pentahydrate tetrachloride | |
| 61442 | |
| 98% | |
| Powder or Chunks |
| 10026-06-9 | |
| Cl4H10O5Sn | |
| MFCD00149864 | |
| tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o | |
| O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4] | |
| 350.59 | |
| 350.6g/mol | |
| Poly Bottle | |
| Tin(IV) Chloride Pentahydrate |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314
DOTInformation : 8/PGIII
RTECSNumber : XP8870000