accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OClSnClClCl

CAS RN 10026-06-9

IUPAC Name: tin(4+) pentahydrate tetrachloride
Synonyms: tin(4+) pentahydrate tetrachloride
Molecular Weight (g/mol): 350.59
Molecular Formula: Cl4H10O5Sn
InChi Key: KHMOASUYFVRATF-UHFFFAOYSA-J
SMILES: O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
Molecular Weight (g/mol) 
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)