missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride hydrate, 98%

Catalog No. p-4407997
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1157022 100 g
AA1157036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1157022 Supplier Thermo Scientific Chemicals Supplier No. 01157022
Only null left

CAS: 10026-06-9 | Cl4Sn | 260.51 g/mol

Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-06-9
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
MDL Number MFCD00149864
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Melting Point 56°C
Physical Form Lump
Quantity 100 g
Assay Percent Range 98%
Solubility Information Soluble in water and alcohol.
Formula Weight 260.50 (Anhydrous)
Chemical Name or Material Tin(IV) chloride hydrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-588-9
RTECSNumber XP8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only