missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride bis(2,4-pentanedionate), 95%

Catalog No. p-7049875
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3053209 10 g
AA3053218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3053209 Supplier Thermo Scientific Chemicals Supplier No. 03053209
Only null left

CAS: 16919-46-3 | C10H14Cl2O4Sn | 387.83 g/mol

It is used as a raw material in synthesis. It is also used as NMR shift reagents and as catalysts for organic synthesis, and precursors to industrial hydroformylation catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16919-46-3
Molecular Formula C10H14Cl2O4Sn
Molecular Weight (g/mol) 387.83
MDL Number MFCD00015316
InChI Key DTMARTPDQMQTRX-VGKOASNMSA-J
Synonym Bis(acetylacetonato)dichlorotin(IV); Tin(IV) bis(acetylacetonate) chloride
SMILES [Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 199°C
Physical Form Crystalline
Quantity 10 g
UN Number UN3146
Solubility Information Soluble in organic solvents.
IUPAC Name tin(4+) bis((2Z)-4-oxopent-2-en-2-olate) dichloride
Formula Weight 387.83
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material Tin(IV) chloride bis(2,4-pentanedionate)
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 240-972-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only