accessibility menu, dialog, popup

UserName

CAS RN 16919-46-3

OOOOSnH3CH3CCH3CH3ClCl

CAS RN 16919-46-3

IUPAC Name: tin(4+) bis((2Z)-4-oxopent-2-en-2-olate) dichloride
Synonyms: tin(4+) bis((2Z)-4-oxopent-2-en-2-olate) dichloride
Molecular Weight (g/mol): 387.83
Molecular Formula: C10H14Cl2O4Sn
InChi Key: DTMARTPDQMQTRX-VGKOASNMSA-J
SMILES: [Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)