missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, anhydrous, 98% (metals basis)

Catalog No. p-7048823
Change view
Click to view available options
Quantity:
500 g
4 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1157136 500 g
AA1157157 4 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1157136 Supplier Thermo Scientific Chemicals Supplier No. 01157136
Only null left

CAS: 7646-78-8 | Cl4Sn | 260.51 g/mol

Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which find applications as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in the presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
MDL Number MFCD00011242
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Boiling Point 114°C
Melting Point -33°C
Color Colorless
Physical Form Liquid
Quantity 500 g
Linear Formula SnCl4
UN Number UN1827
Merck Index 14,8774
Solubility Information Soluble in alcohol,CCl4,benzene,toluene,chloroform,carbon terachloride,gasoline,carbon disulfide and acetone; Soluble in water with much heat evolution.
Formula Weight 260.5
Percent Purity 98%
Refractive Index 1.512
Sensitivity Moisture sensitive
Density 2.26 g/mL
Assay (metals basis)
Chemical Name or Material Tin(IV) chloride, Anhydrous
Show More Show Less
Hazard Category H314-H330-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: STANNIC CHLORIDE, ANHYDROUS
EINECSNumber 231-588-9
RTECSNumber XP8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only