Learn More
Tin(IV) chloride, 99%, anhydrous
CAS: 7646-78-8 | Cl4Sn | 260.52 g/mol
Supplier: Thermo Scientific Chemicals 197510010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpécifications
| 114.0°C | |
| -33.0°C | |
| Brown-Red | |
| 1 L | |
| Cl4Sn | |
| 01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 11,522; 12,486; 13,14; 14,304; 15,311; 17,335 | |
| tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
| HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
| tetrachlorostannane | |
| 24287 | |
| 99% | |
| 2.22 |
| Tin(IV) chloride | |
| 7646-78-8 | |
| Solution | |
| 98.5 to 101.5% (Argentometry) | |
| SnCl4 | |
| 15,8901 | |
| Solubility in water: reacts. Other solubilities: soluble in carbon disulfide, alcohol, carbon, tetrachloride, benzene, toluene, acetone, kerosene, and gasoline | |
| Cl[Sn](Cl)(Cl)Cl | |
| 260.52 | |
| 260.52 | |
| Glass bottle | |
| 2.2200g/mL |
Identifiants chimiques
| 7646-78-8 | |
| 260.52 | |
| tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
| tetrachlorostannane |
| Cl4Sn | |
| HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
| 24287 | |
| Cl[Sn](Cl)(Cl)Cl |
Sécurité et manipulation
GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-588-9
Recommended Storage : Product may develop some turbidity or precipitate
RUO – Research Use Only