missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) 2,3-Naphthalocyanine Dichloride, TCI America™
Supplier: TCI America T29401G
Specifications
| Tin(IV) 2,3-Naphthalocyanine Dichloride | |
| 26857-61-4 | |
| 1 g | |
| MFCD00171716 | |
| Dichloro(2,3-naphthalocyanato)tin(IV) |
| Green | |
| Crystalline Powder | |
| C48H24Cl2N8Sn | |
| 3146 | |
| 902.39 |
Chemical Identifiers
| 26857-61-4 | |
| C48H24Cl2N8Sn | |
| MFCD00171716 | |
| Dichloro(2,3-naphthalocyanato)tin(IV) |
Safety and Handling
TSCA : No