missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) 2,3-Naphthalocyanine Dichloride, TCI America™
$236.80 - $770.07
Chemical Identifiers
| CAS | 26857-61-4 |
|---|---|
| Molecular Formula | C48H24Cl2N8Sn |
| MDL Number | MFCD00171716 |
| Synonym | Dichloro(2,3-naphthalocyanato)tin(IV) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2940200MG
|
TCI America
T2940200MG |
200 mg |
Each for $236.80
|
|
|||||
|
T29401G
|
TCI America
T29401G |
1 g |
Each for $770.07
|
|
|||||
Chemical Identifiers
| 26857-61-4 | |
| C48H24Cl2N8Sn | |
| MFCD00171716 | |
| Dichloro(2,3-naphthalocyanato)tin(IV) |
Specifications
| Green | |
| Crystalline Powder | |
| C48H24Cl2N8Sn | |
| 3146 | |
| 902.39 |
| 26857-61-4 | |
| 200 mg | |
| MFCD00171716 | |
| Dichloro(2,3-naphthalocyanato)tin(IV) | |
| Tin(IV) 2,3-Naphthalocyanine Dichloride |
Safety and Handling
TSCA : No