missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) tetrafluoroborate, 50% w/w aq. soln.

Catalog No. p-7080523
Change view
Click to view available options
Quantity:
1 kg
3 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA89669A1 1 kg
AA89669D5 3 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA89669A1 Supplier Thermo Scientific Chemicals Supplier No. 089669A1
Only null left

CAS: 13814-97-6 | B2F8Sn | 292.32 g/mol

Tin(II) tetrafluoroborate is used in electroplating. It is also used for plating bath tinned copper, tin and tin alloy. Further, it acts as a catalyst for various organic reactions. It is also employed in metal plating. In addition to this, it is used as a petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13814-97-6
Molecular Formula B2F8Sn
Molecular Weight (g/mol) 292.32
MDL Number MFCD00054047
InChI Key JPZSNSDMGTYJIW-UHFFFAOYSA-N
Synonym tin ii fluoroborate, tin fluoroborate, tin 2+ ditetrafluoroborate, tin bis tetrafluoroborate, tin borofluoride, tin fluoborate, tin ii borofluoride, stannous tetrafluoroborate, tin ii tetrafluoroborate
PubChem CID 159698
IUPAC Name tin(2+);ditetrafluoroborate
SMILES [Sn++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 kg
Linear Formula Sn(BF4)2
UN Number UN3264
Solubility Information Miscible with water.
Formula Weight 292.3
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Density 1.670 g/mL
Chemical Name or Material Tin (II) tetrafluoroborate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 237-487-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only