CAS RN 13814-97-6
IUPAC Name:
λ²-tin(2+) bis(tetrafluoroboranuide)
Synonyms:
tin(II) tetrafluoroborate
Molecular Weight (g/mol):
292.32
Molecular Formula:
B2F8Sn
InChi Key:
JPZSNSDMGTYJIW-UHFFFAOYSA-N
SMILES:
[Sn++].F[B-](F)(F)F.F[B-](F)(F)F
1
–
1
of
1
results
1
Tin(II) tetrafluoroborate, 50% w/w aq. soln.
CAS: 13814-97-6 Molecular Formula: B2F8Sn Molecular Weight (g/mol): 292.32 MDL Number: MFCD00054047 InChI Key: JPZSNSDMGTYJIW-UHFFFAOYSA-N Synonym: tin ii fluoroborate,tin fluoroborate,tin 2+ ditetrafluoroborate,tin bis tetrafluoroborate,tin borofluoride,tin fluoborate,tin ii borofluoride,stannous tetrafluoroborate,tin ii tetrafluoroborate PubChem CID: 159698 IUPAC Name: tin(2+);ditetrafluoroborate SMILES: [Sn++].F[B-](F)(F)F.F[B-](F)(F)F