missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) tartrate hydrate, 95%

Catalog No. AA3960414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3960414 25 g
AA3960422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3960414 Supplier Thermo Scientific Chemicals Supplier No. 03960414
Only null left

CAS: 815-85-0 | C4H4O6Sn | 266.78 g/mol

It is used for dyeing and printing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 815-85-0
Molecular Formula C4H4O6Sn
Molecular Weight (g/mol) 266.78
MDL Number MFCD00054358
InChI Key YXTDAZMTQFUZHK-UHFFFAOYNA-L
Synonym unii-6js9u5611x, stannous tartrate, tin tartrate, tartaric acid, tin 2+ salt 1:1, r-r*,r*-2,3-dihydroxybutanedioic acid, tin r-r*,r*-tartrate, butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1
PubChem CID 66545835
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioate;tin(2+)
SMILES [Sn++].OC(C(O)C([O-])=O)C([O-])=O

Specifications

Physical Form Powder
Quantity 25 g
Merck Index 14,8792
Solubility Information slightly soluble in water.
Formula Weight 266.76 (Anhydrous)
Percent Purity 95%
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Tin(II) tartrate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-427-1
RTECSNumber EJ9892500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only