missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) iodide, ultra dry, 99.999% (metals basis)

Catalog No. p-4782188
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1447606 5 g
AA1447614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1447606 Supplier Thermo Scientific Chemicals Supplier No. 01447606
Only null left

CAS: 10294-70-9 | I2Sn | 372.519 g/mol

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3 which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-70-9
Molecular Formula I2Sn
Molecular Weight (g/mol) 372.519
MDL Number MFCD00049545
InChI Key JTDNNCYXCFHBGG-UHFFFAOYSA-L
Synonym tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 25138
IUPAC Name diiodotin
SMILES [Sn](I)I

Specifications

Boiling Point 720°C (decomposition)
Melting Point 320°C
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.99%, metal basis
Solubility Information Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides.
Formula Weight 372.5
Sensitivity Air and moisture sensitive
Density 5.28
Chemical Name or Material Tin(II) Iodide, Ampouled under argon
Hazard Category H314-H335
Hazard Statement When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 233-667-3
RTECSNumber XQ3650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only