accessibility menu, dialog, popup

UserName

CAS RN 10294-70-9

SnII

CAS RN 10294-70-9

IUPAC Name: λ²-tin(2+) diiodide
Synonyms: tin(II) iodide
Molecular Weight (g/mol): 372.52
Molecular Formula: I2Sn
InChi Key: JTDNNCYXCFHBGG-UHFFFAOYSA-L
SMILES: [Sn++].[I-].[I-]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)