missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, For AAS, ≥96.0% (RT), MilliporeSigma™ Supelco™

Catalog No. p-7193496
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019665 50 g
111018458 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019665 Supplier Merck Emd Millipore Supplier No. 9652750G
Only null left

Tin(II) chloride dehydrate converts conjugated dioxolones to aldehyde. It helps as a catalyst in multi-component condensation reaction from aromatic aldehydes, 2-aminopyridines and isonitriles which results in reasonable yield and reducing the reaction time.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
MDL Number MFCD00149863
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
Synonym Stannous chloride dihydrate
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

Linear Formula SnCl2 · 2H2O
Melting Point 37°C to 38°C (decomposition)
Boiling Point 652°C
Quantity 50 g
Formula Weight 225.65
Percent Purity ≥96.0% (RT)
Grade AAS
RTECSNumber XP8850000