missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin AA Standard, 1000 ppm Sn in 15% HCl, Ricca Chemical
$111.50 - $111.50
Chemical Identifiers
| CAS | 7440-31-5 |
|---|---|
| Molecular Formula | Sn |
| Molecular Weight (g/mol) | 118.71 |
| MDL Number | MFCD00133862 |
| InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
| PubChem CID | 5352426 |
| ChEBI | CHEBI:32990 |
| IUPAC Name | tin |
| SMILES | [Sn] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
ASN1KH-100
|
Ricca Chemical Company
ASN1KH100 |
100 mL | Natural LDPE Bottle |
Each for $111.50
|
|
||||
Chemical Identifiers
| 7440-31-5 | |
| 118.71 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| CHEBI:32990 | |
| [Sn] |
| Sn | |
| MFCD00133862 | |
| 5352426 | |
| tin |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Sn | |
| 86.0,15.48,0.09 | |
| Liquid | |
| MFCD00133862 | |
| Approximately 0°C | |
| [Sn] | |
| 118.71 | |
| 5352426 | |
| Tin AA Standard |
| 7440-31-5,7647-01-0,7732-18-5 | |
| 83.0,14.87,0.08 | |
| 1 mL = 1 mg Sn | |
| Sn | |
| Colorless | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| tin | |
| 100 mL | |
| CHEBI:32990 |