missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TIMP2 ELISA Kit

ELISA

$898.00

Specifications

Accession Number P16035
Assay Range 1.6 to 1200 pg/mL
Assay Sensitivity 2 pg/mL
Calibration Range 25 pg/mL to 7000 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHTIMP2
Encompass
View Documents Promotion Details
Invitrogen™
EHTIMP2
96 Tests
Pack for $898.00
Only null left
 
Description

Description

The Human Tissue Inhibitor of MetalloProteinases 2 (TIMP-2) ELISA quantitates Hu TIMP-2 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TIMP-2. Principle of the method The Human TIMP-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
Specifications

Specifications

P16035
2 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
CSC-21 K,DDC8
<10
HRP
Other Research
2°C to 8°C
TIMP2
4 hrs 45 mins
1.6 to 1200 pg/mL
25 pg/mL to 7000 pg/mL
HRP
Proteases, Peptidases & Inhibitors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL- 8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, G-CSF, GM-CSF, IFN-gamma, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, PDGF, SCF, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
7077
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
CSC-21 K, DDC8
1 hrs 20 mins
Videos
SDS
Documents

Documents