missing translation for 'onlineSavingsMsg'
Learn More

Medisca Timolol Maleate USP

Catalog No. 14185436
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185436 1 g
14185407 100 g
14185392 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14185436 Supplier Medisca Supplier No. 036606

Timolol Maleate USP is a white to practically white, odorless or practically odorless powder.

Timolol Maleate USP is a white to practically white, odorless or practically odorless powder.

Specifications

Chemical Name or Material Timolol Maleate USP
Packaging Bottle, 15mL, Amber Glass Wide Mouth Packer, 28-400
CAS 26921-17-5
Type Drug Standard
Molecular Weight (g/mol) 432.4952
Quantity 1 g
Percent Purity 98 to 102 %
Grade USP