missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Timolol Maleate USP
Description
Timolol Maleate USP is a white to practically white, odorless or practically odorless powder.
Specifications
Specifications
| Chemical Name or Material | Timolol Maleate USP |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 26921-17-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 432.4952 |
| Quantity | 100 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |