missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, BV421, Clone: 7D3, BD Horizon™
Mouse Monoclonal Antibody
$861.67 - $861.67
Specifications
| Antigen | TIM-3 (CD366) |
|---|---|
| Clone | 7D3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.Specifications
| TIM-3 (CD366) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human TIM-3 | |
| Primary | |
| Vol. Per test: 5μL |
| 7D3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. |
For Research Use Only.