missing translation for 'onlineSavingsMsg'
Learn More

Tigecycline 98.0+%, TCI America™

Catalog No. T3589100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3589100MG 100 mg
T3589500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3589100MG Supplier TCI America Supplier No. T3589100MG
Only null left

Chemical Identifiers

CAS 220620-09-7
Molecular Formula C29H39N5O8
Molecular Weight (g/mol) 585.66
MDL Number MFCD00935753
InChI Key ZXGBRIBPJBHLMO-SBMFAFPZSA-N
Synonym 9-tert-Butylglycylamidominocycline
PubChem CID 54686904
IUPAC Name N-[(5aR,6aS,7S,9Z,10aS)-9-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-1,10a,12-trihydroxy-8,10,11-trioxo-5,5a,6,6a,7,8,9,10,10a,11-decahydrotetracen-2-yl]-2-(tert-butylamino)acetamide
SMILES CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=C(NC(=O)CNC(C)(C)C)C(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Melting Point 185°C
pH 7.7 - 8.2
Quantity 100 mg
UN Number 3077
Formula Weight 585.66
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tigecycline
RTECSNumber QI7619500
TSCA No
Recommended Storage Freezer