accessibility menu, dialog, popup

UserName

H3CNCH3(S)(S)(R)NHOHNCH3H3CCH3OHO(S)OHOH2NOHOOHNCH3H3C(Z)

CAS RN 220620-09-7

IUPAC Name: N-[(5aR,6aS,7S,9Z,10aS)-9-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-1,10a,12-trihydroxy-8,10,11-trioxo-5,5a,6,6a,7,8,9,10,10a,11-decahydrotetracen-2-yl]-2-(tert-butylamino)acetamide
Synonyms: tygacil way-gar-936 gar-936,tigecycline
Molecular Weight (g/mol): 585.66
Molecular Formula: C29H39N5O8
InChi Key: ZXGBRIBPJBHLMO-SBMFAFPZSA-N
SMILES: CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=C(NC(=O)CNC(C)(C)C)C(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)