missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIGD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101929 Supplier Invitrogen™ Supplier No. PA5101929
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIGD1.

The protein encoded by this gene belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.

Specifications

Antigen TIGD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIGD1
Gene Accession No. Q96MW7
Gene Alias EEYORE; TIGD1; tigger transposable element derived 1; tigger transposable element-derived protein 1
Gene Symbols TIGD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIGD1(Accession Q96MW7), corresponding to amino acid residues Y259-R309.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200765
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less