missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticlopidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T311025G
Specifications
| Ticlopidine Hydrochloride | |
| 210°C | |
| C14H15Cl2NS | |
| 25 g | |
| MTKNGOHFNXIVOS-UHFFFAOYSA-N | |
| hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride | |
| 65335 | |
| 300.24 | |
| Crystalline Powder |
| 53885-35-1 | |
| White | |
| MFCD00079606 | |
| ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf | |
| [H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2 | |
| 300.24 | |
| CHEBI:9589 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 53885-35-1 | |
| 300.24 | |
| MTKNGOHFNXIVOS-UHFFFAOYSA-N | |
| 65335 | |
| hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride |
| C14H15Cl2NS | |
| MFCD00079606 | |
| ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf | |
| CHEBI:9589 | |
| [H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2 |
Safety and Handling
RTECSNumber : XJ9089100
TSCA : No