missing translation for 'onlineSavingsMsg'
Learn More

Tiaramide Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137623
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T317625MG 25 mg
T3176100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T317625MG Supplier TCI America Supplier No. T317625MG
Only null left

Chemical Identifiers

CAS 35941-71-0
Molecular Formula C15H21Cl2N3O3S
Molecular Weight (g/mol) 394.31
MDL Number MFCD01702832
InChI Key XJTXBQFPQSKSPV-UHFFFAOYNA-N
Synonym 5-Chloro-3-[[4-(2-hydroxyethyl)piperazin-1-yl]carbonylmethyl]-2-benzothiazolinone Hydrochloride
PubChem CID 443949
IUPAC Name hydrogen 5-chloro-3-{2-[4-(2-hydroxyethyl)piperazin-1-yl]-2-oxoethyl}-2,3,3a,7a-tetrahydro-1,3-benzothiazol-2-one chloride
SMILES [H+].[Cl-].OCCN1CCN(CC1)C(=O)CN1C2C=C(Cl)C=CC2SC1=O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 25 mg
Formula Weight 392.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tiaramide Hydrochloride
RTECSNumber TL7050000
TSCA No