missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiapride Hydrochloride 99.0+%, TCI America™
Supplier: TCI America T36001G
Specifications
| Tiapride Hydrochloride | |
| White | |
| MFCD00133861 | |
| N-[2-(Diethylamino)ethyl]-2-methoxy-5-(methylsulfonyl)benzamide Hydrochloride | |
| CCN(CC)CCNC(=O)C1=C(C=CC(=C1)S(=O)(=O)C)OC.Cl | |
| 364.885 | |
| 364.89 | |
| Crystalline Powder |
| 51012-33-0 | |
| C15H25ClN2O4S | |
| 1 g | |
| OTFDPNXIVHBTKW-UHFFFAOYSA-N | |
| N-[2-(diethylamino)ethyl]-2-methoxy-5-methylsulfonylbenzamide;hydrochloride | |
| 5702214 | |
| ≥99.0% (HPLC,T) |
Chemical Identifiers
| 51012-33-0 | |
| 364.885 | |
| OTFDPNXIVHBTKW-UHFFFAOYSA-N | |
| 5702214 | |
| CCN(CC)CCNC(=O)C1=C(C=CC(=C1)S(=O)(=O)C)OC.Cl |
| C15H25ClN2O4S | |
| MFCD00133861 | |
| N-[2-(Diethylamino)ethyl]-2-methoxy-5-(methylsulfonyl)benzamide Hydrochloride | |
| N-[2-(diethylamino)ethyl]-2-methoxy-5-methylsulfonylbenzamide;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator