missing translation for 'onlineSavingsMsg'
Learn More

Tiapride Hydrochloride 99.0+%, TCI America™

Catalog No. p-7137812
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3600250MG 250 mg
T36001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3600250MG Supplier TCI America Supplier No. T3600250MG
Only null left

Chemical Identifiers

CAS 51012-33-0
Molecular Formula C15H25ClN2O4S
Molecular Weight (g/mol) 364.885
MDL Number MFCD00133861
InChI Key OTFDPNXIVHBTKW-UHFFFAOYSA-N
Synonym N-[2-(Diethylamino)ethyl]-2-methoxy-5-(methylsulfonyl)benzamide Hydrochloride
PubChem CID 5702214
IUPAC Name N-[2-(diethylamino)ethyl]-2-methoxy-5-methylsulfonylbenzamide;hydrochloride
SMILES CCN(CC)CCNC(=O)C1=C(C=CC(=C1)S(=O)(=O)C)OC.Cl

Specifications

Color White
Quantity 250 mg
Formula Weight 364.89
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tiapride Hydrochloride
TSCA No
Recommended Storage Refrigerator