missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tians epic™ High Performance Static Dissipative Cleanroom Frocks
Lint-free film coated frock for use in critical, ESD, and low particulate clean environments.
Supplier: Tians 846851S
Description
- Lint-free film coated frock
- Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas
- Features include a snap front, elastic wrists
- Mandarin snap collar, no pockets
- Fabric is constructed from a blend of polyethylene and polypropylene
- Finished fabric contains a proprietary additive making it static dissipative
- For use in ISO 6 Cleanroom Class environments
- Neatly folded and cleanroom packed
- 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case
Specifications
| Controlled Environments Frock | |
| Polypropylene, Polyethylene | |
| Single-use | |
| Snaps | |
| Mandarin | |
| 27.9 cm | |
| Cleanroom Packed | |
| 43.1 cm |
| Small | |
| White | |
| Non-sterile | |
| Elastic | |
| 11 in. | |
| Class 1 CPSC 1610, IEST-RP-CC003.3 | |
| 17 in. | |
| Class 1000 (ISO 6) |