missing translation for 'onlineSavingsMsg'
Learn More

Tians epic™ High Performance Static Dissipative Cleanroom Frocks

Catalog No. p-4246902
Change view
Click to view available options
Size:
Small
Medium
X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191300519B Medium
191300519D X-Large
191300519A Small
191300519F 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191300519B Supplier Tians Supplier No. 846851 M
Only null left

Lint-free film coated frock for use in critical, ESD, and low particulate clean environments.

  • Lint-free film coated frock
  • Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas
  • Features include a snap front, elastic wrists
  • Mandarin snap collar, no pockets
  • Fabric is constructed from a blend of polyethylene and polypropylene
  • Finished fabric contains a proprietary additive making it static dissipative
  • For use in ISO 6 Cleanroom Class environments
  • Neatly folded and cleanroom packed
  • 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case

Specifications

Product Type Controlled Environments Frock
Size Medium
Material Polypropylene, Polyethylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Mandarin
Size (English) Chest 11 in.
Size (Metric) Chest 27.9 cm
Certifications/Compliance Class 1 CPSC 1610, IEST-RP-CC003.3
Packaging Type Cleanroom Packed
Length (English) 17 in.
Length (Metric) 43.1 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less