missing translation for 'onlineSavingsMsg'
Learn More

Tianeptine Sodium Salt Hydrate 98.0+%, TCI America™

Supplier:  TCI America T31311G

Quantity 1 g

 View more versions of this product

Catalog No. T31311G


Only null left

Description

Description

Specifications

Chemical Identifiers

30123-17-2
458.933
ZLBSUOGMZDXYKE-UHFFFAOYSA-M
23663953
CN1C2=CC=CC=C2C(C3=C(S1(=O)=O)C=C(C=C3)Cl)NCCCCCCC(=O)[O-].[Na+]
C21H24ClN2NaO4S
MFCD00879851
tianeptine sodium, tianeptine sodium salt, stablon, tatinol, coaxil, +-tianeptine monosodium salt, tianeptine sodium, +, sodium 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoate s,s-dioxide, 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoic acid s,s-dioxide sodium salt
sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-yl)amino]heptanoate

Specifications

Tianeptine Sodium Salt Hydrate
184°C
C21H24ClN2NaO4S
MFCD00879851
ZLBSUOGMZDXYKE-UHFFFAOYSA-M
sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-yl)amino]heptanoate
23663953
≥98.0% (HPLC,N)
30123-17-2
White
1 g
tianeptine sodium, tianeptine sodium salt, stablon, tatinol, coaxil, +-tianeptine monosodium salt, tianeptine sodium, +, sodium 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoate s,s-dioxide, 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoic acid s,s-dioxide sodium salt
CN1C2=CC=CC=C2C(C3=C(S1(=O)=O)C=C(C=C3)Cl)NCCCCCCC(=O)[O-].[Na+]
458.933
458.93
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions