missing translation for 'onlineSavingsMsg'
Learn More

Tianeptine Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. p-7137596
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3131200MG 200 mg
T31311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3131200MG Supplier TCI America Supplier No. T3131200MG

Chemical Identifiers

CAS 30123-17-2
Molecular Formula C21H24ClN2NaO4S
Molecular Weight (g/mol) 458.933
MDL Number MFCD00879851
InChI Key ZLBSUOGMZDXYKE-UHFFFAOYSA-M
Synonym tianeptine sodium, tianeptine sodium salt, stablon, tatinol, coaxil, +-tianeptine monosodium salt, tianeptine sodium, +, sodium 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoate s,s-dioxide, 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoic acid s,s-dioxide sodium salt
PubChem CID 23663953
IUPAC Name sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-yl)amino]heptanoate
SMILES CN1C2=CC=CC=C2C(C3=C(S1(=O)=O)C=C(C=C3)Cl)NCCCCCCC(=O)[O-].[Na+]

Specifications

Melting Point 184°C
Color White
Quantity 200 mg
Formula Weight 458.93
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tianeptine Sodium Salt Hydrate