missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thyrotropin-Releasing Hormone

Catalog No. AAJ62120MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62120MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62120MF Supplier Thermo Scientific Chemicals Supplier No. J62120MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

362.38 g/mol

Two-chain glycoprotein hormone. Activates adenylate cyclase in the thyroid gland, stimulating iodine uptake, thyroxine synthesis and release

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Molecular Weight (g/mol) 362.38
Quantity 50 mg
Packaging Plastic bottle
Form Powder

RUO – Research Use Only