missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Thyroid Stimulating Hormone ELISA Kit DFS Item

ELISA

$874.00 - $874.00

Specifications

Accession Number P01215
Assay Range 2.744 to 2000 pg/mL
Assay Sensitivity 4 pg/mL
Calibration Range 4 pg/mL to 2000 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
EHTSH DFS Item
View Documents
Invitrogen™
EHTSH
96 Tests
Pack for $874.00
Only null left
 
Description

Description

The Human TSH (CGA) ELISA quantitates Hu TSH in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TSH. Principle of the method The Human TSH solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.
Specifications

Specifications

P01215
4 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
CG-ALPHA,FSHA,GPHA1,GPHa,HCG,LHA,TSHA
<10
HRP
Endocrinology
2°C to 8°C
Thyroid Stimulating Hormone
4 hrs 45 mins
2.744 to 2000 pg/mL
4 pg/mL to 2000 pg/mL
HRP
Hormones & Receptors
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
1081, 7252
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
RP11-156F23.1, CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA
1 hrs 20 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'