missing translation for 'onlineSavingsMsg'
Learn More

Thymol Standard, For quantitative NMR, MilliporeSigma™ Supelco™

Catalog No. 111018621
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018621 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018621 Supplier Merck Emd Millipore Supplier No. 504091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

CAS 89-83-8
Product Line TraceCERT
Vapor Pressure 1 mmHg (64°C)
Density 0.965 g/mL (at 25°C)
Linear Formula 2-[(CH3)2 CH]C6H3-5-(CH3)OH
MDL Number MFCD00002309
Melting Point 48°C to 51°C
Boiling Point 232°C
Synonym 2-Isopropyl-5-methylphenol; 5-Methyl-2-(1-methylethyl)phenol; 5-Methyl-2-isopropylphenol; IPMP
Quantity 1 g
Formula Weight 150.22
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC