missing translation for 'onlineSavingsMsg'
Learn More

Thymol Iodide, Powder, Purified, 42%, Spectrum™ Chemical

Catalog No. 18614432
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
5 kg
12 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614432 5 kg Poly Pail
18614431 25 g Amber Glass Bottle
18614428 125 g Amber Glass Bottle
18614430 500 g Amber Glass Bottle
18614429 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614432 Supplier Spectrum Chemical Mfg Cor Supplier No. TH1805KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 552-22-7

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 5 kg
Percent Purity 0.42
Grade Purified

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.