accessibility menu, dialog, popup

UserName

CAS RN 552-22-7

IUPAC Name: 4'-(iodooxy)-2,2'-dimethyl-5,5'-bis(propan-2-yl)-[1,1'-biphenyl]-4-yl hypoiodite
Synonyms: 4'-(iodooxy)-5,5'-diisopropyl-2,2'-dimethyl-[1,1'-biphenyl]-4-yl hypoiodite
Molecular Weight (g/mol): 550.22
Molecular Formula: C20H24I2O2
InChi Key: SHOKWSLXDAIZPP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=C(C)C=C1OI)C1=CC(C(C)C)=C(OI)C=C1C
Boiling Point 
  • (5)
Grade 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)