missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymol Blue sodium salt, ACS reagent
CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol
Supplier: Thermo Scientific Chemicals 420941000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Thymol Blue sodium salt | |
| 62625-21-2 | |
| C27H29NaO5S | |
| OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
| [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O | |
| 488.57 | |
| 488.58 | |
| Glass bottle | |
| Black/Brown or Black/Green to Orange | |
| Powder |
| 283.0°C | |
| Passes Test | |
| MFCD00151093 | |
| Authentic | |
| sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate | |
| 23692293 | |
| ACS Reagent | |
| Acid range: from pH 1.2 (pink) to pH 2.8 (yellow) Alkaline range: from pH 8.0 (yellow) to pH 9.2 (blue) | |
| 100 g |
Chemical Identifiers
| 62625-21-2 | |
| 488.57 | |
| OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
| [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
| C27H29NaO5S | |
| MFCD00151093 | |
| 23692293 |
RUO – Research Use Only