Learn More
Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals
Water soluble pH indicator
$99.58 - $99.58
Chemical Identifiers
| CAS | 62625-21-2 |
|---|---|
| Molecular Formula | C27H29NaO5S |
| Molecular Weight (g/mol) | 488.57 |
| MDL Number | MFCD00151093 |
| InChI Key | OCMIKNBSIRSUPI-FRWNMSGJSA-M |
| PubChem CID | 23692293 |
| IUPAC Name | sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate |
| SMILES | [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Description
Water soluble pH indicatorThymol Blue sodium salt acts as a pH indicator and utilized to indicate the changes in pH. It is involved in the ionization in multi-component drug formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 62625-21-2 | |
| 488.57 | |
| OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
| sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate |
| C27H29NaO5S | |
| MFCD00151093 | |
| 23692293 | |
| [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Specifications
| ∼284°C (decomposition) | |
| C27H29NaO5S | |
| 3861515 | |
| OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
| sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate | |
| 23692293 | |
| ACS | |
| Crystalline |
| 62625-21-2 | |
| MFCD00151093 | |
| Soluble in waterSoluble in 0;1N hydrochloric acid/ethanol. | |
| [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O | |
| 488.57 | |
| 488.58 | |
| 10 g | |
| Thymol Blue sodium salt |
Safety and Handling
H500
EINECSNumber : 263-650-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only