missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue Sodium Salt, ACS Reagent Grade, Ricca Chemical

$143.17 - $143.17

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
RDCT04205F1
View Documents
Ricca Chemical Company
RDCT04205F1
5 g Pouch
Each for $143.17
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62625-21-2
488.57
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
C27H29NaO5S
MFCD00151093
23692293
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

62625-21-2
100.0
MFCD00151093
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
488.57
ACS
Brown to Green
Thymol Blue Sodium Salt
98.0
C27H29NaO5S
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
23692293
Pouch
5 g
Videos
Safety and Handling

Safety and Handling

ShelfLife : 60 months

SDS
Documents

Documents