missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue sodium salt

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol

$75.74 - $1016.82

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
Synonym Thymol Blue, water soluble
PubChem CID 23692293
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2300709
View Documents
Thermo Scientific Chemicals
B2300709
10 g
Each for $75.74
Only null left
 
AAB2300718
View Documents
Thermo Scientific Chemicals
B2300718
50 g
Each for $236.55
Only null left
 
AAB2300730
View Documents
Thermo Scientific Chemicals
B2300730
250 g
Each for $1,016.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62625-21-2
488.57
OCMIKNBSIRSUPI-FRWNMSGJSA-M
23692293
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
C27H29NaO5S
MFCD00151093
Thymol Blue, water soluble
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate

Specifications

∼284°C (decomposition)
C27H29NaO5S
3861515
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
23692293
10 g
62625-21-2
MFCD00151093
Thymol Blue, water soluble
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
488.57
488.58
Thymol Blue sodium salt
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 263-650-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only