missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue sodium salt, 0.04% w/v aq. soln.

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol

$51.90 - $75.03

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38692AE
View Documents
Thermo Scientific Chemicals
038692AE
100 mL
Each for $51.90
Only null left
 
AA38692AP
View Documents
Thermo Scientific Chemicals
038692AP
500 mL
Each for $75.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62625-21-2
488.57
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
C27H29NaO5S
MFCD00151093
23692293
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

62625-21-2
C27H29NaO5S
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
23692293
Odorless
100 mL
Thymol Blue sodium salt
0.04% w/v aq. soln.
MFCD00151093
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
488.57
488.58
0.979 g/mL
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-973-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only